Tisk…

2-[(6-Chloropyridin-3-yl)methyl]-2,3-dihydro-1H-isoindole-1,3-dione 95%

Dodavatel: Apollo Scientific

Synonyms: 2-((6-Chloropyridin-3-yl)methyl)isoindoline-1,3-dione

OR32486-1G OR32486-5G
APOSOR32486-1GEA 9660 CZK
APOSOR32486-1G APOSOR32486-5G
2-[(6-Chloropyridin-3-yl)methyl]-2,3-dihydro-1H-isoindole-1,3-dione 95%
2-[(6-Chloropyridin-3-yl)methyl]-2,3-dihydro-1H-isoindole-1,3-dione
Vzorec: C₁₄H₉ClN₂O₂
MW: 272,686 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09802551
CAS číslo: 120739-60-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR