Tisk…

2-Bromo-N-(2,6-dimethylphenyl)acetamide 95%

Dodavatel: Apollo Scientific

OR32395-1G OR32395-25G OR32395-5G
APOSOR32395-1GEA 5130 CZK
APOSOR32395-1G APOSOR32395-25G APOSOR32395-5G
2-Bromo-N-(2,6-dimethylphenyl)acetamide 95%
2-Bromo-N-(2,6-dimethylphenyl)acetamide
Vzorec: C₁₀H₁₂BrNO
MW: 242,12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00215011
CAS číslo: 32433-61-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR