Tisk…

Ethyl 2-(4-bromobenzamido)acetate

Dodavatel: Apollo Scientific

OR323200-1G OR323200-5G
APOSOR323200-1GEA 0 CZK
APOSOR323200-1G APOSOR323200-5G
Ethyl 2-(4-bromobenzamido)acetate
Ethyl 2-(4-bromobenzamido)acetate
Vzorec: C₁₁H₁₂BrNO₃
MW: 286.122 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00701884
CAS číslo: 207864-21-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR