Tisk…

4-(Azepan-1-yl)benzaldehyde

Dodavatel: Apollo Scientific

OR322936-1G OR322936-500MG
APOSOR322936-1GEA 20830 CZK
APOSOR322936-1G APOSOR322936-500MG
4-(Azepan-1-yl)benzaldehyde
4-(Azepan-1-yl)benzaldehyde
Vzorec: C₁₃H₁₇NO
MW: 203,28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06357359
CAS číslo: 50333-45-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR