4-Bromo-2-(4-methylpiperazin-1-yl)benzonitrile
Dodavatel: Apollo Scientific
OR322769-500MG
OR322769-1G
APOSOR322769-500MGEA
12200
CZK
APOSOR322769-500MG
APOSOR322769-1G
4-Bromo-2-(4-methylpiperazin-1-yl)benzonitrile
4-Bromo-2-(4-methylpiperazin-1-yl)benzonitrile
Vzorec:
C₁₂H₁₄BrN₃ MW: 280.164 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11037544 CAS číslo: 1206969-07-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...