Tisk…

5-Bromo-7-methoxybenzofuran-2-carboxylic acid

Dodavatel: Apollo Scientific
Varování

OR322337-250MG OR322337-1G
APOSOR322337-250MGEA 1290 CZK
APOSOR322337-250MG APOSOR322337-1G
5-Bromo-7-methoxybenzofuran-2-carboxylic acid
5-Bromo-7-methoxybenzofuran-2-carboxylic acid
Vzorec: C₁₀H₇BrO₄
MW: 271,064 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04352513
CAS číslo: 20037-37-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR