Tisk…

N-[(3,5-Dimethyl-1,2-oxazol-4-yl)methyl]aniline 95%

Dodavatel: Apollo Scientific

OR32214-1G
APOSOR32214-1GEA 9660 CZK
APOSOR32214-1G
N-[(3,5-Dimethyl-1,2-oxazol-4-yl)methyl]aniline 95%
N-[(3,5-Dimethyl-1,2-oxazol-4-yl)methyl]aniline
Vzorec: C₁₂H₁₄N₂O
MW: 202,252 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04124300
CAS číslo: 733796-14-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR