2,3,6,7-Tetrahydro-8-hydroxy-1h,5h-benzo[ij]quinolizine-9-carbaldehyde
Dodavatel: Apollo Scientific
OR322086-5G
OR322086-25G
APOSOR322086-5GEA
4570
CZK
APOSOR322086-5G
APOSOR322086-25G
2,3,6,7-Tetrahydro-8-hydroxy-1h,5h-benzo[ij]quinolizine-9-carbaldehyde
2,3,6,7-Tetrahydro-8-hydroxy-1h,5h-benzo[ij]quinolizine-9-carbaldehyde
Vzorec:
C₁₃H₁₅NO₂ Storage Temperature: Ambient |
MDL Number:
MFCD00192477 CAS číslo: 105847-59-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...