Tisk…

2-Benzoyl-1H-imidazole 95%

Dodavatel: Apollo Scientific

OR32196-1G
APOSOR32196-1GEA 6390 CZK
APOSOR32196-1G
2-Benzoyl-1H-imidazole 95%
2-Benzoyl-1H-imidazole
Vzorec: C₁₀H₈N₂O
MW: 172,183 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00707818
CAS číslo: 38353-02-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR