Tisk…

(S)-3-(4-Bromophenyl)piperidine hydrochloride

Dodavatel: Apollo Scientific

OR321550-1G OR321550-250MG
APOSOR321550-1GEA 12730 CZK
APOSOR321550-1G APOSOR321550-250MG
(S)-3-(4-Bromophenyl)piperidine hydrochloride
(S)-3-(4-Bromophenyl)piperidine hydrochloride
Vzorec: C₁₁H₁₄BrN·ClH
MW: 276,601 g/mol
Storage Temperature: Refrigerator
CAS číslo: 2141967-71-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR