Tisk…

Rel-Methyl (3R,6R)-6-methylpiperidine-3-carboxylate hydrochloride

Dodavatel: Apollo Scientific

OR321549-1G OR321549-250MG
APOSOR321549-1GEA 14190 CZK
APOSOR321549-1G APOSOR321549-250MG
Rel-Methyl (3R,6R)-6-methylpiperidine-3-carboxylate hydrochloride
Rel-Methyl (3R,6R)-6-methylpiperidine-3-carboxylate hydrochloride
Vzorec: C₈H₁₅NO₂·ClH
Storage Temperature: Refrigerator
MDL Number: MFCD30181463
CAS číslo: 1009376-86-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR