2-(Piperidin-3-yl)benzamide hydrochloride
Dodavatel: Apollo Scientific
OR321546-1G
OR321546-250MG
APOSOR321546-1GEA
22750
CZK
APOSOR321546-1G
APOSOR321546-250MG
2-(Piperidin-3-yl)benzamide hydrochloride
2-(Piperidin-3-yl)benzamide hydrochloride
Vzorec:
C₁₂H₁₆N₂O·ClH MW: 240,729 g/mol Storage Temperature: Refrigerator |
CAS číslo:
1654722-99-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...