Tisk…

2-(3-Bromopropyl)thiophene

Dodavatel: Apollo Scientific

OR321540-1G OR321540-250MG
APOSOR321540-1GEA 15940 CZK
APOSOR321540-1G APOSOR321540-250MG
2-(3-Bromopropyl)thiophene
2-(3-Bromopropyl)thiophene
Vzorec: C₇H₉BrS
Storage Temperature: Refrigerator
MDL Number: MFCD11613485
CAS číslo: 30134-51-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR