rac-(R)-2-(1-(((9H-Fluoren-9-yl)methoxy)carbonyl)pyrrolidin-3-yl)acetic acid
Dodavatel: Apollo Scientific
OR321538-1G
OR321538-250MG
APOSOR321538-1GEA
11380
CZK
APOSOR321538-1G
APOSOR321538-250MG
rac-(R)-2-(1-(((9H-Fluoren-9-yl)methoxy)carbonyl)pyrrolidin-3-yl)acetic acid
rac-(R)-2-(1-(((9H-Fluoren-9-yl)methoxy)carbonyl)pyrrolidin-3-yl)acetic acid
Vzorec:
C₂₁H₂₁NO₄ MW: 351.396 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD02179121 CAS číslo: 868523-86-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...