5,6-Dichloropicolinaldehyde
Dodavatel: Apollo Scientific
OR321529-1G
OR321529-100MG
APOSOR321529-1GEA
16340
CZK
APOSOR321529-1G
APOSOR321529-100MG
5,6-Dichloropicolinaldehyde
5,6-Dichloropicolinaldehyde
Vzorec:
C₆H₃Cl₂NO MW: 176 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD16606477 CAS číslo: 408526-50-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...