(S)-3-(2-Bromophenyl)piperidine hydrochloride
Dodavatel: Apollo Scientific
OR321521-1G
OR321521-250MG
APOSOR321521-1GEA
31840
CZK
APOSOR321521-1G
APOSOR321521-250MG
(S)-3-(2-Bromophenyl)piperidine hydrochloride
(S)-3-(2-Bromophenyl)piperidine hydrochloride
Vzorec:
C₁₁H₁₄BrN·ClH MW: 276,601 g/mol Storage Temperature: Refrigerator |
CAS číslo:
2416257-97-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...