Tisk…
4-(3-Nitrophenyl)-1,2,3,6-tetrahydropyridine hydrochloride
Dodavatel: Apollo Scientific
OR321516-1G OR321516-250MG
APOSOR321516-1GEA 29200 CZK
APOSOR321516-1G APOSOR321516-250MG
4-(3-Nitrophenyl)-1,2,3,6-tetrahydropyridine hydrochloride
4-(3-Nitrophenyl)-1,2,3,6-tetrahydropyridine hydrochloride
Vzorec: C₁₁H₁₂N₂O₂·ClH
MW: 240.686 g/mol
Storage Temperature: Refrigerator
CAS číslo: 2409597-54-0