Tisk…

Methyl 3-(1,2,3,6-tetrahydropyridin-4-yl)benzoate hydrochloride

Dodavatel: Apollo Scientific

OR321514-1G OR321514-250MG
APOSOR321514-1GEA 18190 CZK
APOSOR321514-1G APOSOR321514-250MG
Methyl 3-(1,2,3,6-tetrahydropyridin-4-yl)benzoate hydrochloride
Methyl 3-(1,2,3,6-tetrahydropyridin-4-yl)benzoate hydrochloride
Vzorec: C₁₃H₁₅NO₂·ClH
MW: 253.725 g/mol
Storage Temperature: Refrigerator
CAS číslo: 2409597-20-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR