Tisk…
4-(3-Chlorophenyl)-1,2,3,6-tetrahydropyridine hydrochloride
Dodavatel: Apollo Scientific
OR321512-1G OR321512-250MG
APOSOR321512-1GEA 18190 CZK
APOSOR321512-1G APOSOR321512-250MG
4-(3-Chlorophenyl)-1,2,3,6-tetrahydropyridine hydrochloride
4-(3-Chlorophenyl)-1,2,3,6-tetrahydropyridine hydrochloride
Vzorec: C₁₁H₁₂ClN·ClH
MW: 230,134 g/mol
Storage Temperature: Refrigerator
CAS číslo: 143017-59-8