Tisk…

4-(1,2,3,6-Tetrahydropyridin-4-yl)benzonitrile hydrochloride

Dodavatel: Apollo Scientific

OR321510-1G OR321510-250MG
APOSOR321510-1GEA 20470 CZK
APOSOR321510-1G APOSOR321510-250MG
4-(1,2,3,6-Tetrahydropyridin-4-yl)benzonitrile hydrochloride
4-(1,2,3,6-Tetrahydropyridin-4-yl)benzonitrile hydrochloride
Vzorec: C₁₂H₁₂N₂·ClH
MW: 220.698 g/mol
Storage Temperature: Refrigerator
CAS číslo: 437998-31-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR