Tisk…

Rel-tert-Butyl (2R,4R)-4-hydroxy-2-methylpiperidine-1-carboxylate

Dodavatel: Apollo Scientific

Synonyms: cis-tert-Butyl 4-hydroxy-2-methylpiperidine-1-carboxylate

OR321509-1G OR321509-250MG
APOSOR321509-1GEA 11810 CZK
APOSOR321509-1G APOSOR321509-250MG
Rel-tert-Butyl (2R,4R)-4-hydroxy-2-methylpiperidine-1-carboxylate
Rel-tert-Butyl (2R,4R)-4-hydroxy-2-methylpiperidine-1-carboxylate
Vzorec: C₁₁H₂₁NO₃
MW: 215,289 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD23105950
CAS číslo: 152491-42-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR