Tisk…

5-(3-Chloro-5-methoxyphenyl)-1,2,3,6-tetrahydropyridine hydrochloride

Dodavatel: Apollo Scientific

OR321507-1G OR321507-250MG
APOSOR321507-1GEA 15940 CZK
APOSOR321507-1G APOSOR321507-250MG
5-(3-Chloro-5-methoxyphenyl)-1,2,3,6-tetrahydropyridine hydrochloride
5-(3-Chloro-5-methoxyphenyl)-1,2,3,6-tetrahydropyridine hydrochloride
Vzorec: C₁₂H₁₄ClNO·ClH
MW: 260,16 g/mol
Storage Temperature: Refrigerator
CAS číslo: 2409597-32-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR