3-(Piperidin-4-yl)benzamide hydrochloride
Dodavatel: Apollo Scientific
OR321500-1G
OR321500-250MG
APOSOR321500-1GEA
20470
CZK
APOSOR321500-1G
APOSOR321500-250MG
3-(Piperidin-4-yl)benzamide hydrochloride
3-(Piperidin-4-yl)benzamide hydrochloride
Vzorec:
C₁₂H₁₆N₂O·ClH Storage Temperature: Refrigerator |
CAS číslo:
2409597-13-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...