tert-Butyl 4-(4-(aminomethyl)phenyl)piperidine-1-carboxylate
Dodavatel: Apollo Scientific
OR321488-250MG
OR321488-500MG
APOSOR321488-250MGEA
4860
CZK
APOSOR321488-250MG
APOSOR321488-500MG
tert-Butyl 4-(4-(aminomethyl)phenyl)piperidine-1-carboxylate
tert-Butyl 4-(4-(aminomethyl)phenyl)piperidine-1-carboxylate
Vzorec:
C₁₇H₂₆N₂O₂ MW: 290,401 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD09038246 CAS číslo: 1198286-87-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...