Tisk…

4-(Piperidin-2-yl)aniline

Dodavatel: Apollo Scientific

OR321487-1G OR321487-250MG
APOSOR321487-1GEA 18190 CZK
APOSOR321487-1G APOSOR321487-250MG
4-(Piperidin-2-yl)aniline
4-(Piperidin-2-yl)aniline
Vzorec: C₁₁H₁₆N₂
Storage Temperature: Refrigerator
MDL Number: MFCD18702927
CAS číslo: 1023277-38-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR