3-(Piperidin-2-yl)aniline
Dodavatel: Apollo Scientific
OR321486-1G
OR321486-250MG
APOSOR321486-1GEA
18190
CZK
APOSOR321486-1G
APOSOR321486-250MG
3-(Piperidin-2-yl)aniline
3-(Piperidin-2-yl)aniline
Vzorec:
C₁₁H₁₆N₂ Storage Temperature: Refrigerator |
MDL Number:
MFCD18702747 CAS číslo: 1203797-19-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...