Tisk…

1-Benzyl-3-bromopyrrolidine

Dodavatel: Apollo Scientific

OR321460-1G OR321460-250MG OR321460-100MG
APOSOR321460-1GEA 41330 CZK
APOSOR321460-1G APOSOR321460-250MG APOSOR321460-100MG
1-Benzyl-3-bromopyrrolidine
1-Benzyl-3-bromopyrrolidine
Vzorec: C₁₁H₁₄BrN
MW: 240,14 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11226925
CAS číslo: 38042-74-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR