Tisk…
2-(Methoxymethoxy)-2-methylpropan-1-amine hydrochloride 97%
Dodavatel: Apollo Scientific
OR321438-100MG OR321438-1G OR321438-500MG
APOSOR321438-100MGEA 23780 CZK
APOSOR321438-100MG APOSOR321438-1G APOSOR321438-500MG
2-(Methoxymethoxy)-2-methylpropan-1-amine hydrochloride 97%
2-(Methoxymethoxy)-2-methylpropan-1-amine hydrochloride
Vzorec: C₆H₁₅NO₂·ClH
MW: 169,65 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31629925
CAS číslo: 2244083-19-8