tert-Butyl 2-(hydroxymethyl)acrylate 95%
Dodavatel: Apollo Scientific
OR321437-1G
OR321437-5G
APOSOR321437-1GEA
20670
CZK
APOSOR321437-1G
APOSOR321437-5G
tert-Butyl 2-(hydroxymethyl)acrylate 95%
tert-Butyl 2-(hydroxymethyl)acrylate
Vzorec:
C₈H₁₄O₃ MW: 158,195 g/mol Bod varu: 72 °C Storage Temperature: Ambient |
MDL Number:
MFCD30490133 CAS číslo: 121065-74-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...