Tisk…

3-(Piperidin-3-yl)benzoic acid hydrochloride

Dodavatel: Apollo Scientific

OR321423-1G OR321423-250MG
APOSOR321423-1GEA 23840 CZK
APOSOR321423-1G APOSOR321423-250MG
3-(Piperidin-3-yl)benzoic acid hydrochloride
3-(Piperidin-3-yl)benzoic acid hydrochloride
Vzorec: C₁₂H₁₅NO₂·ClH
Storage Temperature: Refrigerator
MDL Number: MFCD08460733
CAS číslo: 2007921-14-2

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR