Tisk…

Ethyl 4-(piperidin-4-yl)benzoate hydrochloride

Dodavatel: Apollo Scientific

OR321405-1G OR321405-250MG
APOSOR321405-1GEA 13660 CZK
APOSOR321405-1G APOSOR321405-250MG
Ethyl 4-(piperidin-4-yl)benzoate hydrochloride
Ethyl 4-(piperidin-4-yl)benzoate hydrochloride
Vzorec: C₁₄H₁₉NO₂·ClH
Storage Temperature: Refrigerator
MDL Number: MFCD29060025
CAS číslo: 1453272-47-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR