Tisk…

(S)-2-Benzylpiperidine hydrochloride

Dodavatel: Apollo Scientific

OR321381-1G OR321381-250MG
APOSOR321381-1GEA 18190 CZK
APOSOR321381-1G APOSOR321381-250MG
(S)-2-Benzylpiperidine hydrochloride
(S)-2-Benzylpiperidine hydrochloride
Vzorec: C₁₂H₁₇N·ClH
Storage Temperature: Refrigerator
MDL Number: MFCD30184774
CAS číslo: 2055172-15-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR