Tisk…

2-(Hydroxymethyl)thiazole-4-carboxylic acid 95%

Dodavatel: Apollo Scientific
Varování

OR321365-1G OR321365-100MG OR321365-250MG OR321365-5G
APOSOR321365-1GEA 12700 CZK
APOSOR321365-1G APOSOR321365-100MG APOSOR321365-250MG APOSOR321365-5G
2-(Hydroxymethyl)thiazole-4-carboxylic acid 95%
2-(Hydroxymethyl)thiazole-4-carboxylic acid
Vzorec: C₅H₅NO₃S
MW: 159,163 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12755802
CAS číslo: 221322-09-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR