1-(Aminomethyl)cyclopropane-1-carboxamide hydrochloride 97%
Dodavatel: Apollo Scientific
|
Varování
|
OR321339-250MG
OR321339-1G
APOSOR321339-250MGEA
12630
CZK
APOSOR321339-250MG
APOSOR321339-1G
1-(Aminomethyl)cyclopropane-1-carboxamide hydrochloride 97%
1-(Aminomethyl)cyclopropane-1-carboxamide hydrochloride
Vzorec:
C₅H₁₀N₂O·ClH Storage Temperature: Ambient |
MDL Number:
MFCD28142443 CAS číslo: 1956355-13-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...