Tisk…

1-(1-Ethyl-4-hydroxy-5-methyl-1H-pyrazol-3-yl)ethan-1-one 97%

Dodavatel: Apollo Scientific
Varování

OR321317-1G OR321317-250MG
APOSOR321317-1GEA 5720 CZK
APOSOR321317-1G APOSOR321317-250MG
1-(1-Ethyl-4-hydroxy-5-methyl-1H-pyrazol-3-yl)ethan-1-one 97%
1-(1-Ethyl-4-hydroxy-5-methyl-1H-pyrazol-3-yl)ethan-1-one
Vzorec: C₈H₁₂N₂O₂
MW: 168,193 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11044344
CAS číslo: 1187732-72-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR