Tisk…

Methyl 4-(piperidin-4-yl)benzoate hydrochloride 97%

Dodavatel: Apollo Scientific
Varování

OR321222-250MG
APOSOR321222-250MGEA 1330 CZK
APOSOR321222-250MG
Methyl 4-(piperidin-4-yl)benzoate hydrochloride 97%
Methyl 4-(piperidin-4-yl)benzoate hydrochloride
Vzorec: C₁₃H₁₇NO₂·ClH
MW: 255,741 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD02179141
CAS číslo: 936130-82-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR