Tisk…

(R)-2-Methyl-2-phenylpiperidine 97%

Dodavatel: Apollo Scientific
Varování

OR321211-100MG OR321211-500MG
APOSOR321211-100MGEA 2480 CZK
APOSOR321211-100MG APOSOR321211-500MG
(R)-2-Methyl-2-phenylpiperidine 97%
(R)-2-Methyl-2-phenylpiperidine
Vzorec: C₁₂H₁₇N
Storage Temperature: Refrigerator
MDL Number: MFCD28355920
CAS číslo: 1272031-55-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR