Tisk…

3-Bromo-5-cyanobenzoic acid methyl ester 97%

Dodavatel: Apollo Scientific

Synonyms: Methyl 3-bromo-5-cyanobenzoate

OR321015-1G
APOSOR321015-1GEA 2530 CZK
APOSOR321015-1G
3-Bromo-5-cyanobenzoic acid methyl ester 97%
3-Bromo-5-cyanobenzoic acid methyl ester
Vzorec: C₉H₆BrNO₂
MW: 240,053 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16620379
CAS číslo: 453566-15-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR