Tisk…

2-Amino-6-methoxybenzonitrile

Dodavatel: Apollo Scientific
Varování

OR321007-1G OR321007-25G OR321007-5G
APOSOR321007-1GEA 705 CZK
APOSOR321007-1G APOSOR321007-25G APOSOR321007-5G
2-Amino-6-methoxybenzonitrile
2-Amino-6-methoxybenzonitrile
Vzorec: C₈H₈N₂O
Storage Temperature: Refrigerator
MDL Number: MFCD09745275
CAS číslo: 1591-37-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR