Tisk…

2-Chloro-N-[4-(pyrrolidin-1-yl)phenyl]acetamide 95%

Dodavatel: Apollo Scientific

OR32041-1G OR32041-5G
APOSOR32041-1GEA 15310 CZK
APOSOR32041-1G APOSOR32041-5G
2-Chloro-N-[4-(pyrrolidin-1-yl)phenyl]acetamide 95%
2-Chloro-N-[4-(pyrrolidin-1-yl)phenyl]acetamide
Vzorec: C₁₂H₁₅ClN₂O
MW: 238,713 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00173016
CAS číslo: 251097-15-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR