Tisk…

3-Boc-6-oxo-3-aza-bicyclo[3.1.1]heptane

Dodavatel: Apollo Scientific

Synonyms: tert-Butyl 6-oxo-3-azabicyclo[3.1.1]heptane-3-carboxylate

OR317281-100MG OR317281-250MG
APOSOR317281-100MGEA 3540 CZK
APOSOR317281-100MG APOSOR317281-250MG
3-Boc-6-oxo-3-aza-bicyclo[3.1.1]heptane
3-Boc-6-oxo-3-aza-bicyclo[3.1.1]heptane
Vzorec: C₁₁H₁₇NO₃
MW: 211,258 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17016750
CAS číslo: 1251013-26-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR