Tisk…

1-Boc-azetidine-3-acetaldehyde

Dodavatel: Apollo Scientific
Varování

Synonyms: tert-Butyl 3-(2-oxoethyl)azetidine-1-carboxylate

OR317195-1G OR317195-250MG
APOSOR317195-1GEA 7610 CZK
APOSOR317195-1G APOSOR317195-250MG
1-Boc-azetidine-3-acetaldehyde
1-Boc-azetidine-3-acetaldehyde
Vzorec: C₁₀H₁₇NO₃
Storage Temperature: Refrigerator
MDL Number: MFCD18374993
CAS číslo: 152537-04-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR