Tisk…

1-Boc-3-(aminomethyl)-3-methylazetidine 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: tert-Butyl 3-(aminomethyl)-3-methylazetidine-1-carboxylate

OR317178-1G OR317178-250MG
APOSOR317178-1GEA 6580 CZK
APOSOR317178-1G APOSOR317178-250MG
1-Boc-3-(aminomethyl)-3-methylazetidine 98%
1-Boc-3-(aminomethyl)-3-methylazetidine
Vzorec: C₁₀H₂₀N₂O₂
MW: 200,278 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12408560
CAS číslo: 1158758-85-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR