Tisk…

Methyl trans-3-hydroxycyclobutanecarboxylate

Dodavatel: Apollo Scientific
Varování

OR317101-1G OR317101-500MG OR317101-250MG
APOSOR317101-1GEA 4400 CZK
APOSOR317101-1G APOSOR317101-500MG APOSOR317101-250MG
Methyl trans-3-hydroxycyclobutanecarboxylate
Methyl trans-3-hydroxycyclobutanecarboxylate
Vzorec: C₆H₁₀O₃
Storage Temperature: Ambient
MDL Number: MFCD18483114
CAS číslo: 63485-51-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR