Tisk…
Methyl 6-oxospiro[3.3]heptane-2-carboxylate 97%
Dodavatel: Apollo Scientific
OR317091-1G OR317091-10G OR317091-5G
APOSOR317091-1GEA 8270 CZK
APOSOR317091-1G APOSOR317091-10G APOSOR317091-5G
Methyl 6-oxospiro[3.3]heptane-2-carboxylate 97%
Methyl 6-oxospiro[3.3]heptane-2-carboxylate
Vzorec: C₉H₁₂O₃
MW: 168,19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20922770
CAS číslo: 1138480-98-4