Tisk…

4-Bromo-5-chlorothiophene-2-carboxylic acid 98%

Dodavatel: Apollo Scientific
Varování

OR31657-1G OR31657-250MG OR31657-5G
APOSOR31657-1GEA 12470 CZK
APOSOR31657-1G APOSOR31657-250MG APOSOR31657-5G
4-Bromo-5-chlorothiophene-2-carboxylic acid 98%
4-Bromo-5-chlorothiophene-2-carboxylic acid
Vzorec: C₅H₂BrClO₂S
MW: 241,49 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00847582
CAS číslo: 60729-37-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR