Tisk…

(6S)-5-Azaspiro[2.4]heptane-6-carboxylic acid, N-BOC protected 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: (6S)-5-(tert-Butoxycarbonyl)-5-azaspiro[2.4]heptane-6-carboxylic acid, (S)-5-(tert-Butoxycarbonyl)-5-azaspiro[2.4]heptane-6-carboxylic acid

OR316088-10G OR316088-25G
APOSOR316088-10GEA 2360 CZK
APOSOR316088-10G APOSOR316088-25G
(6S)-5-Azaspiro[2.4]heptane-6-carboxylic acid, N-BOC protected 98%
(6S)-5-Azaspiro[2.4]heptane-6-carboxylic acid, N-BOC protected
Vzorec: C₁₂H₁₉NO₄
MW: 241,284 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11099673
CAS číslo: 1129634-44-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR