Tisk…

(S)-(-)-α-(N-t-BOC-Amino)-γ-Butyrolactone

Dodavatel: Apollo Scientific

Synonyms: Boc-L-Homoserine lactone, Boc-Hse-Lactone

OR316009-1G OR316009-5G
APOSOR316009-1GEA 1580 CZK
APOSOR316009-1G APOSOR316009-5G
(S)-(-)-α-(N-t-BOC-Amino)-γ-Butyrolactone
(S)-(-)-α-(N-t-BOC-Amino)-γ-Butyrolactone
Vzorec: C₉H₁₅NO₄
Storage Temperature: Ambient
MDL Number: MFCD04973958
CAS číslo: 40856-59-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR