Tisk…

2-(Cyclopropyl)thiophene-3-carboxylic acid 95%

Dodavatel: Apollo Scientific

OR315886-1G OR315886-250MG
APOSOR315886-1GEA 16340 CZK
APOSOR315886-1G APOSOR315886-250MG
2-(Cyclopropyl)thiophene-3-carboxylic acid 95%
2-(Cyclopropyl)thiophene-3-carboxylic acid
Vzorec: C₈H₈O₂S
Storage Temperature: Refrigerator
MDL Number: MFCD32689354
CAS číslo: 2104675-28-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR