Tisk…

Methyl 4-(cyclopropyl)thiophene-2-carboxylate 95%

Dodavatel: Apollo Scientific

OR315881-1G OR315881-250MG
APOSOR315881-1GEA 16340 CZK
APOSOR315881-1G APOSOR315881-250MG
Methyl 4-(cyclopropyl)thiophene-2-carboxylate 95%
Methyl 4-(cyclopropyl)thiophene-2-carboxylate
Vzorec: C₉H₁₀O₂S
MW: 182,24 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD32689351
CAS číslo: 2111296-73-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR